missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trimethylsilylmethyl)allyl acetate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 381380050

 View more versions of this product

Catalog No. AC381380050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-(Trimethylsilylmethyl)allyl acetate
97.5
0.8770g/mL
71°C
97.5% min. (GC)
C9H18O2Si
CH3CO2CH2C(=CH2)CH2Si(CH3)3
5g
2-trimethylsilylmethyl allyl acetate, 2-trimethylsilyl methyl allyl acetate, 2-acetoxymethyl-3-trimethylsilyl propene, 2-trimethylsilyl methyl-2-propen-1-yl acetate, 2-trimethylsilyl methyl prop-2-en-1-yl acetate, acmc-1bfax, 2-trimethylsilylmethyl prop-2-enyl acetate, 2-acetoxymethyl allyl-trimethylsilane
IKQWABMHZKGCLX-UHFFFAOYSA-N
2-(trimethylsilylmethyl)prop-2-enyl acetate
4428347
98%
72047-94-0
100.0
95°C (7.0mmHg)
Authentic
Glass bottle
1.4380 to 1.44
MFCD00075170
0.877
Solubility in water: reacts
CC(=O)OCC(=C)C[Si](C)(C)C
186.33
186.23

Chemical Identifiers

72047-94-0
186.33
IKQWABMHZKGCLX-UHFFFAOYSA-N
4428347
CC(=O)OCC(=C)C[Si](C)(C)C
C9H18O2Si
MFCD00075170
2-trimethylsilylmethyl allyl acetate, 2-trimethylsilyl methyl allyl acetate, 2-acetoxymethyl-3-trimethylsilyl propene, 2-trimethylsilyl methyl-2-propen-1-yl acetate, 2-trimethylsilyl methyl prop-2-en-1-yl acetate, acmc-1bfax, 2-trimethylsilylmethyl prop-2-enyl acetate, 2-acetoxymethyl allyl-trimethylsilane
2-(trimethylsilylmethyl)prop-2-enyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Reacts violently with water.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only