Learn More
2-(Trimethylsilylmethyl)allyl acetate, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 72047-94-0 |
---|---|
Molecular Formula | C9H18O2Si |
Molecular Weight (g/mol) | 186.33 |
MDL Number | MFCD00075170 |
InChI Key | IKQWABMHZKGCLX-UHFFFAOYSA-N |
Synonym | 2-trimethylsilylmethyl allyl acetate, 2-trimethylsilyl methyl allyl acetate, 2-acetoxymethyl-3-trimethylsilyl propene, 2-trimethylsilyl methyl-2-propen-1-yl acetate, 2-trimethylsilyl methyl prop-2-en-1-yl acetate, acmc-1bfax, 2-trimethylsilylmethyl prop-2-enyl acetate, 2-acetoxymethyl allyl-trimethylsilane |
PubChem CID | 4428347 |
IUPAC Name | 2-[(trimethylsilyl)methyl]prop-2-en-1-yl acetate |
SMILES | CC(=O)OCC(=C)C[Si](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC381380010
|
Thermo Scientific Chemicals
381380010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC381380050
|
Thermo Scientific Chemicals
381380050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC381380250
|
Thermo Scientific Chemicals
381380250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
72047-94-0 | |
186.33 | |
IKQWABMHZKGCLX-UHFFFAOYSA-N | |
4428347 | |
CC(=O)OCC(=C)C[Si](C)(C)C |
C9H18O2Si | |
MFCD00075170 | |
2-trimethylsilylmethyl allyl acetate, 2-trimethylsilyl methyl allyl acetate, 2-acetoxymethyl-3-trimethylsilyl propene, 2-trimethylsilyl methyl-2-propen-1-yl acetate, 2-trimethylsilyl methyl prop-2-en-1-yl acetate, acmc-1bfax, 2-trimethylsilylmethyl prop-2-enyl acetate, 2-acetoxymethyl allyl-trimethylsilane | |
2-[(trimethylsilyl)methyl]prop-2-en-1-yl acetate |
Specifications
72047-94-0 | |
100.0 | |
95°C (7.0mmHg) | |
Authentic | |
Glass bottle | |
1.4380 to 1.44 | |
MFCD00075170 | |
0.877 | |
Solubility in water: reacts | |
CC(=O)OCC(=C)C[Si](C)(C)C | |
186.33 | |
186.23 | |
2-(Trimethylsilylmethyl)allyl acetate |
97.5 | |
0.8770g/mL | |
71°C | |
97.5% min. (GC) | |
C9H18O2Si | |
CH3CO2CH2C(=CH2)CH2Si(CH3)3 | |
1g | |
2-trimethylsilylmethyl allyl acetate, 2-trimethylsilyl methyl allyl acetate, 2-acetoxymethyl-3-trimethylsilyl propene, 2-trimethylsilyl methyl-2-propen-1-yl acetate, 2-trimethylsilyl methyl prop-2-en-1-yl acetate, acmc-1bfax, 2-trimethylsilylmethyl prop-2-enyl acetate, 2-acetoxymethyl allyl-trimethylsilane | |
IKQWABMHZKGCLX-UHFFFAOYSA-N | |
2-[(trimethylsilyl)methyl]prop-2-en-1-yl acetate | |
4428347 | |
98% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Reacts violently with water.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only