missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ 2000 mL Bottles for Sorvall BIOS 16 Bioprocessing Centrifuge
Thermo Scientific™ 2000 mL Bottle designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
$705.36 - $1626.77
Catalog Number | Mfr. No. | Sterility | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sterility | Material | Price | Quantity | ||||
75-003-872
|
Thermo Scientific™
75003872 |
Non-sterile | Polypropylene |
Pack for $705.36
|
|
||||
75003871
|
Thermo Scientific™
75003871 |
Sterile | Polypropylene |
Pack of 2 Each for $958.31
|
|
||||
75-003-870
|
Thermo Scientific™
75003870 |
Non-sterile | PPCO |
Pack of 2 Each for $1,626.77
|
|
||||
Description
Process a 16L harvest in a single run with the Thermo Scientific™ 2000 mL Bottle, the ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
The Thermo Scientific™ 2000 mL Bottles are ideal solution for the separation of large volume harvests. Designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge.Specifications
2000 mL | |
White |