missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H63420MC

 View more versions of this product

Catalog No. AAH63420MC


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Colorless to Yellow
>110°C (230°F)
1.572
100 mg
2r,5r-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, r,r-me-rajphos, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C
264.305
264.3
Liquid
1044256-04-3
1.105 g/mL
C15H21O2P
MFCD09842713
Air and moisture sensitive
JAKXTLMNIHPLEU-VXGBXAGGSA-N
2-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
71310562
97%

Chemical Identifiers

1044256-04-3
264.305
JAKXTLMNIHPLEU-VXGBXAGGSA-N
71310562
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C
C15H21O2P
MFCD09842713
2r,5r-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, r,r-me-rajphos, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min
2-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only