missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%, Thermo Scientific Chemicals
$144.81 - $483.93
Chemical Identifiers
CAS | 1044256-04-3 |
---|---|
Molecular Formula | C15H21O2P |
Molecular Weight (g/mol) | 264.305 |
MDL Number | MFCD09842713 |
InChI Key | JAKXTLMNIHPLEU-VXGBXAGGSA-N |
Synonym | 2r,5r-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, r,r-me-rajphos, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min |
PubChem CID | 71310562 |
IUPAC Name | 2-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane |
SMILES | CC1CCC(P1C2=CC=CC=C2C3OCCO3)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH63420MC
|
Thermo Scientific Chemicals
H63420MC |
100 mg |
Each for $144.81
|
|
||||||
AAH63420ME
|
Thermo Scientific Chemicals
H63420ME |
500 mg |
Each for $483.93
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1044256-04-3 | |
264.305 | |
JAKXTLMNIHPLEU-VXGBXAGGSA-N | |
71310562 | |
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C |
C15H21O2P | |
MFCD09842713 | |
2r,5r-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, r,r-me-rajphos, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min | |
2-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane |
Specifications
1044256-04-3 | |
1.105 g/mL | |
C15H21O2P | |
MFCD09842713 | |
Air and moisture sensitive | |
JAKXTLMNIHPLEU-VXGBXAGGSA-N | |
2-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane | |
71310562 | |
97% | |
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane |
Colorless to Yellow | |
>110°C (230°F) | |
1.572 | |
100 mg | |
2r,5r-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, r,r-me-rajphos, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min | |
CC1CCC(P1C2=CC=CC=C2C3OCCO3)C | |
264.305 | |
264.3 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only