missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC60614DA

 View more versions of this product

Catalog No. CC60614DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

3-(1-Methyl-1H-pyrazol-3-yl)aniline
95%
C10H11N3
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole
CN1C=CC(=N1)C2=CC(=CC=C2)N
173.219
173.22
175202-37-6
Amber Glass Bottle
1g
ZRXOZSROZJGRRH-UHFFFAOYSA-N
3-(1-methylpyrazol-3-yl)aniline
2735396
97%

Chemical Identifiers

175202-37-6
173.219
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole
3-(1-methylpyrazol-3-yl)aniline
C10H11N3
ZRXOZSROZJGRRH-UHFFFAOYSA-N
2735396
CN1C=CC(=N1)C2=CC(=CC=C2)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions