missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Thermo Scientific™
Chemical Identifiers
CAS | 175202-37-6 |
---|---|
Molecular Formula | C10H11N3 |
Molecular Weight (g/mol) | 173.219 |
InChI Key | ZRXOZSROZJGRRH-UHFFFAOYSA-N |
Synonym | 3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole |
PubChem CID | 2735396 |
IUPAC Name | 3-(1-methylpyrazol-3-yl)aniline |
SMILES | CN1C=CC(=N1)C2=CC(=CC=C2)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC60614CB
|
Thermo Scientific Chemicals
CC60614CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC60614DA
|
Thermo Scientific Chemicals
CC60614DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC60614DE
|
Thermo Scientific Chemicals
CC60614DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
175202-37-6 | |
173.219 | |
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole | |
3-(1-methylpyrazol-3-yl)aniline |
C10H11N3 | |
ZRXOZSROZJGRRH-UHFFFAOYSA-N | |
2735396 | |
CN1C=CC(=N1)C2=CC(=CC=C2)N |
Specifications
175202-37-6 | |
Amber Glass Bottle | |
250mg | |
ZRXOZSROZJGRRH-UHFFFAOYSA-N | |
3-(1-methylpyrazol-3-yl)aniline | |
2735396 | |
97% |
95% | |
C10H11N3 | |
3-1-methyl-1h-pyrazol-3-yl aniline, 3-1-methylpyrazol-3-yl aniline, 3-1-methyl-1h-pyrazol-3-yl-phenylamine, benzenamine, 3-1-methyl-1h-pyrazol-3-yl, 3-1-methylpyrazol-3-yl phenylamine, 3-1-methyl-1h-pyrazol-3-yl phenylamine, 3-3-aminophenyl-1-methyl-1h-pyrazole | |
CN1C=CC(=N1)C2=CC(=CC=C2)N | |
173.219 | |
173.22 | |
3-(1-Methyl-1H-pyrazol-3-yl)aniline |
Safety and Handling
GHS Signal Word: Warning