missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3,3-Trifluoro-2,2-dimethylpropionic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H26053MD

 View more versions of this product

Catalog No. AAH26053MD


Explore available promotions

Description

Description

3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications

Specifications

3,3,3-Trifluoro-2,2-dimethylpropionic acid
66°C to 73°C
C5H7F3O2
250 mg
3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid
CC(C)(C(=O)O)C(F)(F)F
156.104
156.12
889940-13-0
76°C to 77°C (14 mmHg)
MFCD08445819
UN3261
VMFSJVUPIXOCFO-UHFFFAOYSA-N
3,3,3-trifluoro-2,2-dimethylpropanoic acid
19867367
97%

Chemical Identifiers

889940-13-0
156.104
VMFSJVUPIXOCFO-UHFFFAOYSA-N
19867367
CC(C)(C(=O)O)C(F)(F)F
C5H7F3O2
MFCD08445819
3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid
3,3,3-trifluoro-2,2-dimethylpropanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only