Learn More
3,3,3-Trifluoro-2,2-dimethylpropionic acid, 97%, Thermo Scientific Chemicals
$205.82 - $577.51
Chemical Identifiers
CAS | 889940-13-0 |
---|---|
Molecular Formula | C5H7F3O2 |
Molecular Weight (g/mol) | 156.104 |
MDL Number | MFCD08445819 |
InChI Key | VMFSJVUPIXOCFO-UHFFFAOYSA-N |
Synonym | 3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid |
PubChem CID | 19867367 |
IUPAC Name | 3,3,3-trifluoro-2,2-dimethylpropanoic acid |
SMILES | CC(C)(C(=O)O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH26053MD
|
Thermo Scientific Chemicals
H26053MD |
250 mg |
Each for $205.82
|
|
|||||
AAH2605303
|
Thermo Scientific Chemicals
H2605303 |
1 g |
Each for $577.51
|
|
|||||
Description
3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Chemical Identifiers
889940-13-0 | |
156.104 | |
VMFSJVUPIXOCFO-UHFFFAOYSA-N | |
19867367 | |
CC(C)(C(=O)O)C(F)(F)F |
C5H7F3O2 | |
MFCD08445819 | |
3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid | |
3,3,3-trifluoro-2,2-dimethylpropanoic acid |
Specifications
889940-13-0 | |
76°C to 77°C (14 mmHg) | |
MFCD08445819 | |
UN3261 | |
VMFSJVUPIXOCFO-UHFFFAOYSA-N | |
3,3,3-trifluoro-2,2-dimethylpropanoic acid | |
19867367 | |
97% |
66°C to 73°C | |
C5H7F3O2 | |
250 mg | |
3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid | |
CC(C)(C(=O)O)C(F)(F)F | |
156.104 | |
156.12 | |
3,3,3-Trifluoro-2,2-dimethylpropionic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only