missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3-Dimethyl-1-butanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3019418

 View more versions of this product

Catalog No. AAH3019418

Explore available promotions

Description

Description

3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

3,3-Dimethyl-1-butanol
-60°C
142°C to 143°C
Alcohol-like
1.414
MFCD00002928
UN1987
Slightly soluble in water.
CC(C)(C)CCO
102.18
102.18
624-95-3
0.81
56°C (133°F)
C6H14O
(CH3)3CCH2CH2OH
50 g
3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096
DUXCSEISVMREAX-UHFFFAOYSA-N
3,3-dimethylbutan-1-ol
12233
97%

Chemical Identifiers

624-95-3
102.18
DUXCSEISVMREAX-UHFFFAOYSA-N
12233
CC(C)(C)CCO
C6H14O
MFCD00002928
3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096
3,3-dimethylbutan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 210-872-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only