Learn More
3,3-Dimethyl-1-butanol, 97%
CAS: 624-95-3 | C6H14O | 102.18 g/mol
$211.84 - $729.07
Chemical Identifiers
| CAS | 624-95-3 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.18 |
| MDL Number | MFCD00002928 |
| InChI Key | DUXCSEISVMREAX-UHFFFAOYSA-N |
| Synonym | 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096 |
| PubChem CID | 12233 |
| IUPAC Name | 3,3-dimethylbutan-1-ol |
| SMILES | CC(C)(C)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3019409
|
Thermo Scientific Chemicals
H3019409 |
10 g |
N/A
|
|
|||||
|
AAH3019418
|
Thermo Scientific Chemicals
H3019418 |
50 g |
N/A
|
|
|||||
Description
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 624-95-3 | |
| 102.18 | |
| DUXCSEISVMREAX-UHFFFAOYSA-N | |
| 12233 | |
| CC(C)(C)CCO |
| C6H14O | |
| MFCD00002928 | |
| 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096 | |
| 3,3-dimethylbutan-1-ol |
Specifications
| 624-95-3 | |
| 0.81 | |
| 56°C (133°F) | |
| C6H14O | |
| (CH3)3CCH2CH2OH | |
| 10 g | |
| 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096 | |
| DUXCSEISVMREAX-UHFFFAOYSA-N | |
| 3,3-dimethylbutan-1-ol | |
| 12233 | |
| 97% |
| -60°C | |
| 142°C to 143°C | |
| Alcohol-like | |
| 1.414 | |
| MFCD00002928 | |
| UN1987 | |
| Slightly soluble in water. | |
| CC(C)(C)CCO | |
| 102.18 | |
| 102.18 | |
| 3,3-Dimethyl-1-butanol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 210-872-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only