missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic Acid 98.0+%, TCI America™

Supplier:  TCI America T31625G

 View more versions of this product

Catalog No. T31625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic Acid
210°C
C13H17BO4
5 g
OPWAPCOSDAFWFB-UHFFFAOYSA-N
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
2734653
≥98.0% (GC,T)
269409-73-6
White
MFCD03411930
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O
248.09
248.09
Crystalline Powder

Chemical Identifiers

269409-73-6
248.09
OPWAPCOSDAFWFB-UHFFFAOYSA-N
2734653
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O
C13H17BO4
MFCD03411930
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions