missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2062006

 View more versions of this product

Catalog No. AAB2062006


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4,5-Trihydroxybenzamide
∼240°C (decomposition)
MFCD00014800
2644549
RBQIPEJXQPQFJX-UHFFFAOYSA-N
3,4,5-trihydroxybenzamide
69256
98%
618-73-5
C7H7NO4
5 g
gallamide, benzamide, 3,4,5-trihydroxy, gallamid, gallic acid amide, benzamide,4,5-trihydroxy, acmc-1b99c, 3,4,5-trihydroxy benzamide, benzamide,3,4,5-trihydroxy, 3,4,5-tris oxidanyl benzamide, 3,4,5-trihydroxybenzamide
NC(=O)C1=CC(O)=C(O)C(O)=C1
169.14
169.14

Chemical Identifiers

618-73-5
169.14
RBQIPEJXQPQFJX-UHFFFAOYSA-N
69256
NC(=O)C1=CC(O)=C(O)C(O)=C1
C7H7NO4
MFCD00014800
gallamide, benzamide, 3,4,5-trihydroxy, gallamid, gallic acid amide, benzamide,4,5-trihydroxy, acmc-1b99c, 3,4,5-trihydroxy benzamide, benzamide,3,4,5-trihydroxy, 3,4,5-tris oxidanyl benzamide, 3,4,5-trihydroxybenzamide
3,4,5-trihydroxybenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-561-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only