missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals

$75.66 - $229.40

Chemical Identifiers

CAS 618-73-5
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00014800
InChI Key RBQIPEJXQPQFJX-UHFFFAOYSA-N
Synonym gallamide, benzamide, 3,4,5-trihydroxy, gallamid, gallic acid amide, benzamide,4,5-trihydroxy, acmc-1b99c, 3,4,5-trihydroxy benzamide, benzamide,3,4,5-trihydroxy, 3,4,5-tris oxidanyl benzamide, 3,4,5-trihydroxybenzamide
PubChem CID 69256
IUPAC Name 3,4,5-trihydroxybenzamide
SMILES NC(=O)C1=CC(O)=C(O)C(O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2062006
View Documents
Thermo Scientific Chemicals
B2062006
5 g
Each for $75.66
 
AAB2062014
View Documents
Thermo Scientific Chemicals
B2062014
25 g
Each for $229.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

618-73-5
169.14
RBQIPEJXQPQFJX-UHFFFAOYSA-N
69256
NC(=O)C1=CC(O)=C(O)C(O)=C1
C7H7NO4
MFCD00014800
gallamide, benzamide, 3,4,5-trihydroxy, gallamid, gallic acid amide, benzamide,4,5-trihydroxy, acmc-1b99c, 3,4,5-trihydroxy benzamide, benzamide,3,4,5-trihydroxy, 3,4,5-tris oxidanyl benzamide, 3,4,5-trihydroxybenzamide
3,4,5-trihydroxybenzamide

Specifications

618-73-5
C7H7NO4
5 g
gallamide, benzamide, 3,4,5-trihydroxy, gallamid, gallic acid amide, benzamide,4,5-trihydroxy, acmc-1b99c, 3,4,5-trihydroxy benzamide, benzamide,3,4,5-trihydroxy, 3,4,5-tris oxidanyl benzamide, 3,4,5-trihydroxybenzamide
NC(=O)C1=CC(O)=C(O)C(O)=C1
169.14
169.14
3,4,5-Trihydroxybenzamide
∼240°C (decomposition)
MFCD00014800
2644549
RBQIPEJXQPQFJX-UHFFFAOYSA-N
3,4,5-trihydroxybenzamide
69256
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-561-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only