Learn More
3,4,7,8-Tetramethyl-1,10-phenanthroline, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 172220010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,4,7,8-Tetramethyl-1,10-phenanthroline | |
277.0°C to 280.0°C | |
Authentic | |
Glass bottle | |
1 g | |
Solubility in water: insoluble. Other solubilities: soluble in 1,4-dioxane,acetone,dichloromethane,,n,n-dimethylformamide,n,n-dimethylacetamide,meth,anol | |
CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C | |
236.32 | |
236.32 | |
Fine Crystalline Powder |
1660-93-1 | |
Beige | |
98+% | |
C16H16N2 | |
1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline | |
NPAXPTHCUCUHPT-UHFFFAOYSA-N | |
3,4,7,8-tetramethyl-1,10-phenanthroline | |
74265 | |
98+% |
Chemical Identifiers
1660-93-1 | |
236.32 | |
1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline | |
3,4,7,8-tetramethyl-1,10-phenanthroline |
C16H16N2 | |
NPAXPTHCUCUHPT-UHFFFAOYSA-N | |
74265 | |
CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C |
Safety and Handling
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician
EINECSNumber : 216-762-4
RUO – Research Use Only