missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,7,8-Tetramethyl-1,10-phenanthroline, 98+%, Thermo Scientific Chemicals

$175.11 - $648.25

Chemical Identifiers

CAS 1660-93-1
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.32
InChI Key NPAXPTHCUCUHPT-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline
PubChem CID 74265
IUPAC Name 3,4,7,8-tetramethyl-1,10-phenanthroline
SMILES CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172220010
SDS View Documents
Thermo Scientific Chemicals
172220010
1 g Glass bottle
Each for $175.11
 
AC172220050
SDS View Documents
Thermo Scientific Chemicals
172220050
5 g Glass bottle
Each for $648.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1660-93-1
236.32
1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline
3,4,7,8-tetramethyl-1,10-phenanthroline
C16H16N2
NPAXPTHCUCUHPT-UHFFFAOYSA-N
74265
CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C

Specifications

1660-93-1
Beige
98+%
C16H16N2
1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline
NPAXPTHCUCUHPT-UHFFFAOYSA-N
3,4,7,8-tetramethyl-1,10-phenanthroline
74265
98+%
3,4,7,8-Tetramethyl-1,10-phenanthroline
277.0°C to 280.0°C
Authentic
Glass bottle
1 g
Solubility in water: insoluble. Other solubilities: soluble in 1,4-dioxane,acetone,dichloromethane,,n,n-dimethylformamide,n,n-dimethylacetamide,meth,anol
CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C
236.32
236.32
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician

EINECSNumber : 216-762-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only