missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(4-Aminophenyl)propionic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 2393-17-1 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
InChI Key | WXOHKMNWMKZMND-UHFFFAOYSA-N |
Synonym | 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid |
PubChem CID | 75451 |
IUPAC Name | 3-(4-aminophenyl)propanoic acid |
SMILES | C1=CC(=CC=C1CCC(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC443600010
|
Thermo Scientific Chemicals
443600010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC443600050
|
Thermo Scientific Chemicals
443600050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2393-17-1 | |
165.19 | |
3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid | |
3-(4-aminophenyl)propanoic acid |
C9H11NO2 | |
WXOHKMNWMKZMND-UHFFFAOYSA-N | |
75451 | |
C1=CC(=CC=C1CCC(=O)O)N |
Specifications
2393-17-1 | |
96% min. (HPLC) | |
C9H11NO2 | |
3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid | |
C1=CC(=CC=C1CCC(=O)O)N | |
165.19 | |
165.19 | |
3-(4-Aminophenyl)propionic acid |
Authentic | |
Glass Bottle | |
1g | |
WXOHKMNWMKZMND-UHFFFAOYSA-N | |
3-(4-aminophenyl)propanoic acid | |
75451 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 219-245-1
RUO â Research Use Only