missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Chlorophenoxy)-1,2-propanediol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1738622

 View more versions of this product

Catalog No. AAA1738622


Explore available promotions

Description

Description

It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

Solubility
Slightly soluble in water.

Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
Specifications

Specifications

3-(4-Chlorophenoxy)-1,2-propanediol
104-29-0
C9H11ClO3
chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
MXOAEAUPQDYUQM-UHFFFAOYSA-N
3-(4-chlorophenoxy)propane-1,2-diol
7697
202.64
78°C to 82°C
100 g
MFCD00021990
Slightly soluble in water.
C1=CC(=CC=C1OCC(CO)O)Cl
202.634
CHEBI:3642
99%

Chemical Identifiers

104-29-0
202.634
MXOAEAUPQDYUQM-UHFFFAOYSA-N
7697
3-(4-chlorophenoxy)propane-1,2-diol
C9H11ClO3
MFCD00021990
chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
CHEBI:3642
C1=CC(=CC=C1OCC(CO)O)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 203-192-6

RTECSNumber : TY4260000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only