Learn More
3-(4-Chlorophenoxy)-1,2-propanediol, 99%, Thermo Scientific Chemicals
$87.37 - $969.75
Chemical Identifiers
CAS | 104-29-0 |
---|---|
Molecular Formula | C9H11ClO3 |
Molecular Weight (g/mol) | 202.634 |
MDL Number | MFCD00021990 |
InChI Key | MXOAEAUPQDYUQM-UHFFFAOYSA-N |
Synonym | chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine |
PubChem CID | 7697 |
ChEBI | CHEBI:3642 |
IUPAC Name | 3-(4-chlorophenoxy)propane-1,2-diol |
SMILES | C1=CC(=CC=C1OCC(CO)O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1738614
|
Thermo Scientific Chemicals
A1738614 |
25 g |
Each for $87.37
|
|
|||||
AAA1738622
|
Thermo Scientific Chemicals
A1738622 |
100 g |
Each for $245.53
|
|
|||||
AAA1738636
|
Thermo Scientific Chemicals
A1738636 |
500 g |
Each for $969.75
|
|
|||||
Description
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.
Solubility
Slightly soluble in water.
Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
Chemical Identifiers
104-29-0 | |
202.634 | |
MXOAEAUPQDYUQM-UHFFFAOYSA-N | |
7697 | |
3-(4-chlorophenoxy)propane-1,2-diol |
C9H11ClO3 | |
MFCD00021990 | |
chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine | |
CHEBI:3642 | |
C1=CC(=CC=C1OCC(CO)O)Cl |
Specifications
78°C to 82°C | |
25 g | |
MFCD00021990 | |
Slightly soluble in water. | |
C1=CC(=CC=C1OCC(CO)O)Cl | |
202.634 | |
CHEBI:3642 | |
99% |
104-29-0 | |
C9H11ClO3 | |
chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine | |
MXOAEAUPQDYUQM-UHFFFAOYSA-N | |
3-(4-chlorophenoxy)propane-1,2-diol | |
7697 | |
202.64 | |
3-(4-Chlorophenoxy)-1,2-propanediol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 203-192-6
RTECSNumber : TY4260000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only