missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1587336

 View more versions of this product

Catalog No. AAA1587336


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,4-Dichlorobenzoic acid
205°C to 208°C
MFCD00002492
2044777
VPHHJAOJUJHJKD-UHFFFAOYSA-N
3,4-dichlorobenzoic acid
5817
191.01
51-44-5
C7H4Cl2O2
500 g
benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
OC(=O)C1=CC=C(Cl)C(Cl)=C1
191.01
CHEBI:49401
99%

Chemical Identifiers

51-44-5
191.01
VPHHJAOJUJHJKD-UHFFFAOYSA-N
5817
3,4-dichlorobenzoic acid
C7H4Cl2O2
MFCD00002492
benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
CHEBI:49401
OC(=O)C1=CC=C(Cl)C(Cl)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-099-2

RTECSNumber : DG7175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only