Learn More
3,4-Dichlorobenzoic acid, 99%, Thermo Scientific Chemicals
$49.52 - $565.92
Chemical Identifiers
CAS | 51-44-5 |
---|---|
Molecular Formula | C7H4Cl2O2 |
Molecular Weight (g/mol) | 191.01 |
MDL Number | MFCD00002492 |
InChI Key | VPHHJAOJUJHJKD-UHFFFAOYSA-N |
Synonym | benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609 |
PubChem CID | 5817 |
ChEBI | CHEBI:49401 |
IUPAC Name | 3,4-dichlorobenzoic acid |
SMILES | OC(=O)C1=CC=C(Cl)C(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1587314
|
Thermo Scientific Chemicals
A1587314 |
25 g |
Each for $49.52
|
|
|||||
AAA1587322
|
Thermo Scientific Chemicals
A1587322 |
100 g |
Each for $126.92
|
|
|||||
AAA1587336
|
Thermo Scientific Chemicals
A1587336 |
500 g |
Each for $565.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51-44-5 | |
191.01 | |
VPHHJAOJUJHJKD-UHFFFAOYSA-N | |
5817 | |
3,4-dichlorobenzoic acid |
C7H4Cl2O2 | |
MFCD00002492 | |
benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609 | |
CHEBI:49401 | |
OC(=O)C1=CC=C(Cl)C(Cl)=C1 |
Specifications
51-44-5 | |
C7H4Cl2O2 | |
25 g | |
benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609 | |
OC(=O)C1=CC=C(Cl)C(Cl)=C1 | |
191.01 | |
CHEBI:49401 | |
99% |
205°C to 208°C | |
MFCD00002492 | |
2044777 | |
VPHHJAOJUJHJKD-UHFFFAOYSA-N | |
3,4-dichlorobenzoic acid | |
5817 | |
191.01 | |
3,4-Dichlorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-099-2
RTECSNumber : DG7175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only