missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1206906

 View more versions of this product

Catalog No. AAA1206906

Explore available promotions

Description

Description

It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

3- (4-Hydroxy-3-methoxyphenyl)propionic acid
87°C to 92°C
MFCD00016558
2110370
Soluble in dimethyl sulfoxide and methanol.
COC1=C(C=CC(=C1)CCC(=O)O)O
196.202
CHEBI:86612
97%
1135-23-5
C10H12O4
5 g
3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid
BOLQJTPHPSDZHR-UHFFFAOYSA-N
3-(4-hydroxy-3-methoxyphenyl)propanoic acid
14340
196.2

Chemical Identifiers

1135-23-5
196.202
BOLQJTPHPSDZHR-UHFFFAOYSA-N
14340
3-(4-hydroxy-3-methoxyphenyl)propanoic acid
C10H12O4
MFCD00016558
3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid
CHEBI:86612
COC1=C(C=CC(=C1)CCC(=O)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 214-489-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only