Learn More
3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 97%, Thermo Scientific Chemicals
$143.26 - $524.92
Chemical Identifiers
CAS | 1135-23-5 |
---|---|
Molecular Formula | C10H12O4 |
Molecular Weight (g/mol) | 196.202 |
MDL Number | MFCD00016558 |
InChI Key | BOLQJTPHPSDZHR-UHFFFAOYSA-N |
Synonym | 3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid |
PubChem CID | 14340 |
ChEBI | CHEBI:86612 |
IUPAC Name | 3-(4-hydroxy-3-methoxyphenyl)propanoic acid |
SMILES | COC1=C(C=CC(=C1)CCC(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1206903
|
Thermo Scientific Chemicals
A1206903 |
1 g |
Each for $143.26
|
|
|||||
AAA1206906
|
Thermo Scientific Chemicals
A1206906 |
5 g |
Each for $524.92
|
|
|||||
Description
It is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a caffeine metabolite which showed high antioxidant activity. It is a very sensitive biomarker for the consumption of relatively small amount of coffee. an be used to inhibit prostaglandin E(2) production.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Chemical Identifiers
1135-23-5 | |
196.202 | |
BOLQJTPHPSDZHR-UHFFFAOYSA-N | |
14340 | |
3-(4-hydroxy-3-methoxyphenyl)propanoic acid |
C10H12O4 | |
MFCD00016558 | |
3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid | |
CHEBI:86612 | |
COC1=C(C=CC(=C1)CCC(=O)O)O |
Specifications
1135-23-5 | |
C10H12O4 | |
1 g | |
3-4-hydroxy-3-methoxyphenyl propanoic acid, 3-4-hydroxy-3-methoxyphenyl propionic acid, dihydroferulic acid, hydroferulic acid, dihydroconiferylic acid, unii-o01rnc700m, benzenepropanoic acid, 4-hydroxy-3-methoxy, 3-4-hydroxymethyl propionic acid, .beta.-4-hydroxy-3-methoxyphenyl propionic acid, 3-4-hydroxy-3-methoxy-phenyl propanoic acid | |
BOLQJTPHPSDZHR-UHFFFAOYSA-N | |
3-(4-hydroxy-3-methoxyphenyl)propanoic acid | |
14340 | |
196.2 | |
3- (4-Hydroxy-3-methoxyphenyl)propionic acid |
87°C to 92°C | |
MFCD00016558 | |
2110370 | |
Soluble in dimethyl sulfoxide and methanol. | |
COC1=C(C=CC(=C1)CCC(=O)O)O | |
196.202 | |
CHEBI:86612 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 214-489-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only