missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
3-(4-Methoxyphenyl)propionic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1365514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
3-(4-Methoxyphenyl)propionic acid | |
102°C to 106°C | |
MFCD00002777 | |
1953326 | |
FIUFLISGGHNPSM-UHFFFAOYSA-N | |
3-(4-methoxyphenyl)propanoic acid | |
95750 | |
98% |
1929-29-9 | |
C10H12O3 | |
25 g | |
3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy | |
COC1=CC=C(C=C1)CCC(=O)O | |
180.203 | |
180.2 |
Identifiants chimiques
1929-29-9 | |
180.203 | |
FIUFLISGGHNPSM-UHFFFAOYSA-N | |
95750 | |
COC1=CC=C(C=C1)CCC(=O)O |
C10H12O3 | |
MFCD00002777 | |
3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy | |
3-(4-methoxyphenyl)propanoic acid |
Sécurité et manipulation
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 217-678-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only