missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(4-Methoxyphenyl)propionic acid, 98%, Thermo Scientific Chemicals
$323.55 - $906.52
Chemical Identifiers
CAS | 1929-29-9 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.203 |
MDL Number | MFCD00002777 |
InChI Key | FIUFLISGGHNPSM-UHFFFAOYSA-N |
Synonym | 3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy |
PubChem CID | 95750 |
IUPAC Name | 3-(4-methoxyphenyl)propanoic acid |
SMILES | COC1=CC=C(C=C1)CCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1365514
|
Thermo Scientific Chemicals
A1365514 |
25 g |
Each for $323.55
|
|
|||||
AAA1365522
|
Thermo Scientific Chemicals
A1365522 |
100 g |
Each for $906.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1929-29-9 | |
180.203 | |
FIUFLISGGHNPSM-UHFFFAOYSA-N | |
95750 | |
COC1=CC=C(C=C1)CCC(=O)O |
C10H12O3 | |
MFCD00002777 | |
3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy | |
3-(4-methoxyphenyl)propanoic acid |
Specifications
1929-29-9 | |
C10H12O3 | |
25 g | |
3-4-methoxyphenyl propanoic acid, 3-4-methoxyphenyl propionic acid, benzenepropanoic acid, 4-methoxy, p-methoxyhydrocinnamic acid, 3-p-methoxyphenyl propionic acid, 3-4-methoxy-phenyl-propionic acid, 4-methoxyhydrocinnamic acid, 3-4-methoxyphenyl-propionic acid, 4-methoxyhydro cinnamic acid, hydrocinnamic acid, p-methoxy | |
COC1=CC=C(C=C1)CCC(=O)O | |
180.203 | |
180.2 | |
3-(4-Methoxyphenyl)propionic acid |
102°C to 106°C | |
MFCD00002777 | |
1953326 | |
FIUFLISGGHNPSM-UHFFFAOYSA-N | |
3-(4-methoxyphenyl)propanoic acid | |
95750 | |
98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-678-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only