missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dibromo-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1836114

 View more versions of this product

Catalog No. AAA1836114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dibromo-4-hydroxybenzoic acid
269°C to 273°C
MFCD00002548
2416147
PHWAJJWKNLWZGJ-UHFFFAOYSA-N
3,5-dibromo-4-hydroxybenzoic acid
76857
295.93
3337-62-0
C7H4Br2O3
25 g
bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009
C1=C(C=C(C(=C1Br)O)Br)C(=O)O
295.914
CHEBI:1395
98%

Chemical Identifiers

3337-62-0
295.914
PHWAJJWKNLWZGJ-UHFFFAOYSA-N
76857
3,5-dibromo-4-hydroxybenzoic acid
C7H4Br2O3
MFCD00002548
bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009
CHEBI:1395
C1=C(C=C(C(=C1Br)O)Br)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-075-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only