missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dibromo-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals
$84.18 - $273.93
Chemical Identifiers
CAS | 3337-62-0 |
---|---|
Molecular Formula | C7H4Br2O3 |
Molecular Weight (g/mol) | 295.914 |
MDL Number | MFCD00002548 |
InChI Key | PHWAJJWKNLWZGJ-UHFFFAOYSA-N |
Synonym | bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009 |
PubChem CID | 76857 |
ChEBI | CHEBI:1395 |
IUPAC Name | 3,5-dibromo-4-hydroxybenzoic acid |
SMILES | C1=C(C=C(C(=C1Br)O)Br)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1836114
|
Thermo Scientific Chemicals
A1836114 |
25 g |
N/A
|
|
||||||
AAA1836122
|
Thermo Scientific Chemicals
A1836122 |
100 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3337-62-0 | |
295.914 | |
PHWAJJWKNLWZGJ-UHFFFAOYSA-N | |
76857 | |
3,5-dibromo-4-hydroxybenzoic acid |
C7H4Br2O3 | |
MFCD00002548 | |
bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009 | |
CHEBI:1395 | |
C1=C(C=C(C(=C1Br)O)Br)C(=O)O |
Specifications
3337-62-0 | |
C7H4Br2O3 | |
25 g | |
bromoxynylbenzoic acid, dbha, benzoic acid, 3,5-dibromo-4-hydroxy, dibrhbz, unii-95z5a9epei, 3,5-dibromo-4-hydroxybenzoate, 3,5-dibromo-4-hydroxy-benzoic acid, 3',5'-dibromo-4-hydroxybenzoic acid, 95z5a9epei, rarechem al bo 0009 | |
C1=C(C=C(C(=C1Br)O)Br)C(=O)O | |
295.914 | |
CHEBI:1395 | |
98% |
269°C to 273°C | |
MFCD00002548 | |
2416147 | |
PHWAJJWKNLWZGJ-UHFFFAOYSA-N | |
3,5-dibromo-4-hydroxybenzoic acid | |
76857 | |
295.93 | |
3,5-Dibromo-4-hydroxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 222-075-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only