missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1073218

 View more versions of this product

Catalog No. AAA1073218


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt
>300°C
MFCD00009798
3640643
NMWCVZCSJHJYFW-UHFFFAOYSA-M
sodium;3,5-dichloro-2-hydroxybenzenesulfonate
3085077
99%
54970-72-8
C6H3Cl2NaO4S
50 g
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]
265.036
265.05

Chemical Identifiers

54970-72-8
265.036
NMWCVZCSJHJYFW-UHFFFAOYSA-M
3085077
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+]
C6H3Cl2NaO4S
MFCD00009798
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt
sodium;3,5-dichloro-2-hydroxybenzenesulfonate
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 259-416-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only