missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1073230
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt | |
>300°C | |
MFCD00009798 | |
3640643 | |
NMWCVZCSJHJYFW-UHFFFAOYSA-M | |
sodium;3,5-dichloro-2-hydroxybenzenesulfonate | |
3085077 | |
99% |
54970-72-8 | |
C6H3Cl2NaO4S | |
250 g | |
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt | |
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+] | |
265.036 | |
265.05 |
Chemical Identifiers
54970-72-8 | |
265.036 | |
NMWCVZCSJHJYFW-UHFFFAOYSA-M | |
3085077 | |
C1=C(C=C(C(=C1Cl)O)S(=O)(=O)[O-])Cl.[Na+] |
C6H3Cl2NaO4S | |
MFCD00009798 | |
sodium 3,5-dichloro-2-hydroxybenzenesulfonate, dhbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid sodium salt, sodium 3,5-dichloro-2-hydroxybenzenesulphonate, dchbs, 3,5-dichloro-2-hydroxybenzenesulfonic acid, sodium salt, 2,4-dichlorophenol-6-sulfonic acid sodium salt, sodium 3,5-chloro-6-hydroxybenzenesulfonate, benzenesulfonic acid, 3,5-dichloro-2-hydroxy-, monosodium salt, 3,5-dichloro-2-hydroxybenzenesulphonic acid sodium salt | |
sodium;3,5-dichloro-2-hydroxybenzenesulfonate |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 259-416-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only