missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1950706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3,5-Difluorobenzenesulfonyl chloride | |
57°C to 59°C | |
C6H3ClF2O2S | |
5 g | |
8053561 | |
3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03 | |
FC1=CC(=CC(F)=C1)S(Cl)(=O)=O | |
212.59 | |
212.6 |
210532-25-5 | |
>110°C (230°F) | |
MFCD02091378 | |
UN3261 | |
Moisture sensitive | |
IIQKIICAOXAXEJ-UHFFFAOYSA-N | |
3,5-difluorobenzenesulfonyl chloride | |
2774058 | |
97% |
Chemical Identifiers
210532-25-5 | |
212.59 | |
IIQKIICAOXAXEJ-UHFFFAOYSA-N | |
2774058 | |
FC1=CC(=CC(F)=C1)S(Cl)(=O)=O |
C6H3ClF2O2S | |
MFCD02091378 | |
3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03 | |
3,5-difluorobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures