Learn More
3,5-Difluorobenzoyl chloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2140014
Description
3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.
Solubility
It reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and bases. Moisture Sensitive
Specifications
3,5-Difluorobenzoyl chloride | |
1.41 | |
69°C (156°F) | |
1.503 | |
25 g | |
4307685 | |
3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride | |
OYZWEOORLJBPMA-UHFFFAOYSA-N | |
3,5-difluorobenzoyl chloride | |
145600 | |
≥98% |
129714-97-2 | |
173°C to 175°C | |
C7H3ClF2O | |
MFCD00010309 | |
UN3265 | |
Moisture sensitive | |
It reacts with water. | |
C1=C(C=C(C=C1F)F)C(=O)Cl | |
176.547 | |
176.55 |
Chemical Identifiers
129714-97-2 | |
176.547 | |
OYZWEOORLJBPMA-UHFFFAOYSA-N | |
145600 | |
C1=C(C=C(C=C1F)F)C(=O)Cl |
C7H3ClF2O | |
MFCD00010309 | |
3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride | |
3,5-difluorobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only