missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Difluorobenzoyl chloride, 98+%, Thermo Scientific Chemicals

$123.81 - $414.55

Chemical Identifiers

CAS 129714-97-2
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD00010309
InChI Key OYZWEOORLJBPMA-UHFFFAOYSA-N
Synonym 3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride
PubChem CID 145600
IUPAC Name 3,5-difluorobenzoyl chloride
SMILES C1=C(C=C(C=C1F)F)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2140006
View Documents
Thermo Scientific Chemicals
B2140006
5 g
Each for $123.81
 
AAB2140014
View Documents
Thermo Scientific Chemicals
B2140014
25 g
Each for $414.55
 
Description

Description

3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluorobenzoyl chloride, is used as an pharmaceutical intermediate, organic intermediate and chemical synthesis.

Solubility
It reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong acids and bases. Moisture Sensitive
Specifications

Chemical Identifiers

129714-97-2
176.547
OYZWEOORLJBPMA-UHFFFAOYSA-N
145600
C1=C(C=C(C=C1F)F)C(=O)Cl
C7H3ClF2O
MFCD00010309
3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride
3,5-difluorobenzoyl chloride

Specifications

129714-97-2
173°C to 175°C
C7H3ClF2O
MFCD00010309
UN3265
Moisture sensitive
It reacts with water.
C1=C(C=C(C=C1F)F)C(=O)Cl
176.547
176.55
3,5-Difluorobenzoyl chloride
1.41
69°C (156°F)
1.503
5 g
4307685
3,5-difluorobenzoylchloride, benzoyl chloride, 3,5-difluoro, 3,5-difluorobenzoic acid chloride, pubchem20777, acmc-1chrc, intermediates-zcf02002, 3,5difluorobenzoyl chloride, 3,5-di-fluorobenzoyl chloride, 3,5-difluoro-benzoyl chloride, 3,5-difluoro-benzoyl-chloride
OYZWEOORLJBPMA-UHFFFAOYSA-N
3,5-difluorobenzoyl chloride
145600
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only