missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3(5)-Phenyl-1H-pyrazole, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 2458-26-6
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00159654
InChI Key OEDUIFSDODUDRK-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
PubChem CID 17155
IUPAC Name 5-phenyl-1H-pyrazole
SMILES N1N=CC=C1C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC296030025
SDS View Documents
Thermo Scientific Chemicals
296030025
2.5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2458-26-6
144.18
OEDUIFSDODUDRK-UHFFFAOYSA-N
17155
N1N=CC=C1C1=CC=CC=C1
C9H8N2
MFCD00159654
3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
5-phenyl-1H-pyrazole

Specifications

2458-26-6
100.0
99% min. (GC)
C9H8N2
2.5g
3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
OEDUIFSDODUDRK-UHFFFAOYSA-N
5-phenyl-1H-pyrazole
17155
99+%
99.0
Authentic
Glass bottle
MFCD00159654
23,177
Solubility in water: .. Other solubilities: halogenated solvents,alcohols
N1N=CC=C1C1=CC=CC=C1
144.18
144.18
3(5)-Phenyl-1H-pyrazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only