missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,6-Difluorophthalic anhydride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H26215MC

 View more versions of this product

Catalog No. AAH26215MC


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,6-Difluorophthalic anhydride
218°C to 221°C
MFCD00134537
Moisture sensitive
AVLRPSLTCCWJKC-UHFFFAOYSA-N
4,7-difluoro-2-benzofuran-1,3-dione
513918
97%
652-40-4
C8H2F2O3
100 mg
3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride
C1=CC(=C2C(=C1F)C(=O)OC2=O)F
184.098
184.1

Chemical Identifiers

652-40-4
184.098
AVLRPSLTCCWJKC-UHFFFAOYSA-N
513918
C1=CC(=C2C(=C1F)C(=O)OC2=O)F
C8H2F2O3
MFCD00134537
3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride
4,7-difluoro-2-benzofuran-1,3-dione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only