missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,6-Difluorophthalic anhydride, 97%, Thermo Scientific Chemicals

$85.63 - $250.23

Chemical Identifiers

CAS 652-40-4
Molecular Formula C8H2F2O3
Molecular Weight (g/mol) 184.098
MDL Number MFCD00134537
InChI Key AVLRPSLTCCWJKC-UHFFFAOYSA-N
Synonym 3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride
PubChem CID 513918
IUPAC Name 4,7-difluoro-2-benzofuran-1,3-dione
SMILES C1=CC(=C2C(=C1F)C(=O)OC2=O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26215MC
View Documents
Thermo Scientific Chemicals
H26215MC
100 mg
Each for $85.63
 
AAH26215ME
View Documents
Thermo Scientific Chemicals
H26215ME
500 mg
Each for $250.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

652-40-4
184.098
AVLRPSLTCCWJKC-UHFFFAOYSA-N
513918
C1=CC(=C2C(=C1F)C(=O)OC2=O)F
C8H2F2O3
MFCD00134537
3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride
4,7-difluoro-2-benzofuran-1,3-dione

Specifications

652-40-4
C8H2F2O3
100 mg
3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride
C1=CC(=C2C(=C1F)C(=O)OC2=O)F
184.098
184.1
3,6-Difluorophthalic anhydride
218°C to 221°C
MFCD00134537
Moisture sensitive
AVLRPSLTCCWJKC-UHFFFAOYSA-N
4,7-difluoro-2-benzofuran-1,3-dione
513918
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only