Learn More
3,7-Dimethyl-3-octanol, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 379065000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 7-Dimethyl-3-octanol | |
97.0 | |
0.8280g/mL | |
76°C | |
97% min. (GC) | |
C10H22O | |
(CH3)2CH(CH2)3C(CH3)(OH)CH2CH3 | |
500mL | |
tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670 | |
DLHQZZUEERVIGQ-UHFFFAOYSA-N | |
3,7-dimethyloctan-3-ol | |
6548 | |
158.28 |
78-69-3 | |
100.0 | |
193.0°C | |
Authentic | |
Glass bottle | |
1.4310 to 1.4350 | |
MFCD00004482 | |
0.828 | |
Solubility in water: .. Other solubilities: soluble in alcohol | |
CCC(C)(CCCC(C)C)O | |
158.28 | |
CHEBI:84242 | |
97+% |
Chemical Identifiers
78-69-3 | |
158.28 | |
DLHQZZUEERVIGQ-UHFFFAOYSA-N | |
6548 | |
3,7-dimethyloctan-3-ol |
C10H22O | |
MFCD00004482 | |
tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670 | |
CHEBI:84242 | |
CCC(C)(CCCC(C)C)O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
GHS Signal Word: Warning
EINECSNumber : 201-133-9
RUO â Research Use Only