missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,7-Dimethyl-3-octanol, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 78-69-3
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00004482
InChI Key DLHQZZUEERVIGQ-UHFFFAOYSA-N
Synonym tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670
PubChem CID 6548
ChEBI CHEBI:84242
IUPAC Name 3,7-dimethyloctan-3-ol
SMILES CCC(C)(CCCC(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC379061000
SDS View Documents
Thermo Scientific Chemicals
379061000
100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC379065000
SDS View Documents
Thermo Scientific Chemicals
379065000
500mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

78-69-3
158.28
DLHQZZUEERVIGQ-UHFFFAOYSA-N
6548
3,7-dimethyloctan-3-ol
C10H22O
MFCD00004482
tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670
CHEBI:84242
CCC(C)(CCCC(C)C)O

Specifications

78-69-3
100.0
193.0°C
Authentic
Glass bottle
1.4310 to 1.4350
MFCD00004482
0.828
Solubility in water: .. Other solubilities: soluble in alcohol
CCC(C)(CCCC(C)C)O
158.28
CHEBI:84242
97+%
97.0
0.8280g/mL
76°C
97% min. (GC)
C10H22O
(CH3)2CH(CH2)3C(CH3)(OH)CH2CH3
100mL
tetrahydrolinalool, 3,7-dimethyl-3-octanol, 3-octanol, 3,7-dimethyl, linalool tetrahydride, 2,6-dimethyl-6-octanol, 3,7-dimethyloctanol-3, fema no. 3060, 1-3,7-dimethyloctan-3-ol, dsstox_cid_9110, dsstox_rid_78670
DLHQZZUEERVIGQ-UHFFFAOYSA-N
3,7-dimethyloctan-3-ol
6548
158.28
3, 7-Dimethyl-3-octanol
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth

GHS Signal Word: Warning

EINECSNumber : 201-133-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only