missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(9-Anthryl)propionic acid, 96%, Thermo Scientific Chemicals
$250.19 - $250.19
Chemical Identifiers
CAS | 41034-83-7 |
---|---|
Molecular Formula | C17H14O2 |
Molecular Weight (g/mol) | 250.30 |
MDL Number | MFCD00068641 |
InChI Key | BDGMYCZUEIGHJH-UHFFFAOYSA-N |
Synonym | 9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid |
PubChem CID | 170457 |
IUPAC Name | 3-anthracen-9-ylpropanoic acid |
SMILES | OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3418403
|
Thermo Scientific Chemicals
H3418403 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
41034-83-7 | |
250.30 | |
BDGMYCZUEIGHJH-UHFFFAOYSA-N | |
170457 | |
OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12 |
C17H14O2 | |
MFCD00068641 | |
9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid | |
3-anthracen-9-ylpropanoic acid |
Specifications
41034-83-7 | |
C17H14O2 | |
1 g | |
BDGMYCZUEIGHJH-UHFFFAOYSA-N | |
3-anthracen-9-ylpropanoic acid | |
170457 | |
96% |
193°C to 197°C | |
MFCD00068641 | |
9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid | |
OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12 | |
250.30 | |
250.3 | |
3-(9-Anthryl)propionic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only