missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Acetamidobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2383303

 View more versions of this product

Catalog No. AAB2383303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Acetamidobenzeneboronic acid
∼263°C (decomposition)
MFCD00236013
3278316
IBTSWKLSEOGJGJ-UHFFFAOYSA-N
(3-acetamidophenyl)boronic acid
157274
98%
78887-39-5
C8H10BNO3
1 g
3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid
B(C1=CC(=CC=C1)NC(=O)C)(O)O
178.982
178.98

Chemical Identifiers

78887-39-5
178.982
IBTSWKLSEOGJGJ-UHFFFAOYSA-N
157274
B(C1=CC(=CC=C1)NC(=O)C)(O)O
C8H10BNO3
MFCD00236013
3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid
(3-acetamidophenyl)boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only