missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Acetamidobenzeneboronic acid, 98%, Thermo Scientific Chemicals
$134.05 - $2392.29
Chemical Identifiers
CAS | 78887-39-5 |
---|---|
Molecular Formula | C8H10BNO3 |
Molecular Weight (g/mol) | 178.982 |
MDL Number | MFCD00236013 |
InChI Key | IBTSWKLSEOGJGJ-UHFFFAOYSA-N |
Synonym | 3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid |
PubChem CID | 157274 |
IUPAC Name | (3-acetamidophenyl)boronic acid |
SMILES | B(C1=CC(=CC=C1)NC(=O)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2383303
|
Thermo Scientific Chemicals
B2383303 |
1 g |
Each for $134.05
|
|
|||||
AAB2383306
|
Thermo Scientific Chemicals
B2383306 |
5 g |
Each for $443.77
|
|
|||||
AAB2383314
|
Thermo Scientific Chemicals
B2383314 |
25 g |
Each for $2,392.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78887-39-5 | |
178.982 | |
IBTSWKLSEOGJGJ-UHFFFAOYSA-N | |
157274 | |
B(C1=CC(=CC=C1)NC(=O)C)(O)O |
C8H10BNO3 | |
MFCD00236013 | |
3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid | |
(3-acetamidophenyl)boronic acid |
Specifications
78887-39-5 | |
C8H10BNO3 | |
1 g | |
3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid, boronic acid, 3-acetylamino phenyl, n-3-dihydroxyboranyl phenyl acetamide, 3-acetylamino phenylboronic acid | |
B(C1=CC(=CC=C1)NC(=O)C)(O)O | |
178.982 | |
178.98 | |
3-Acetamidobenzeneboronic acid |
∼263°C (decomposition) | |
MFCD00236013 | |
3278316 | |
IBTSWKLSEOGJGJ-UHFFFAOYSA-N | |
(3-acetamidophenyl)boronic acid | |
157274 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only