missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Acetylpyridine 98.0+%, TCI America™
Supplier: TCI America A0112250G
Specifications
3-Acetylpyridine | |
Yellow | |
C7H7NO | |
250 g | |
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one | |
CC(=O)C1=CN=CC=C1 | |
121.139 | |
121.14 | |
Liquid |
350-03-8 | |
220°C | |
MFCD00006396 | |
2810 | |
WEGYGNROSJDEIW-UHFFFAOYSA-N | |
1-pyridin-3-ylethanone | |
9589 | |
≥98.0% (GC) |
Chemical Identifiers
350-03-8 | |
121.139 | |
WEGYGNROSJDEIW-UHFFFAOYSA-N | |
9589 | |
CC(=O)C1=CN=CC=C1 |
C7H7NO | |
MFCD00006396 | |
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one | |
1-pyridin-3-ylethanone |
Safety and Handling
EINECSNumber : (5)-3706
RTECSNumber : OB5425000
TSCA : Yes