missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Acetylpyridine 98.0+%, TCI America™

$90.86 - $475.97

Chemical Identifiers

CAS 350-03-8
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006396
InChI Key WEGYGNROSJDEIW-UHFFFAOYSA-N
Synonym 3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
PubChem CID 9589
IUPAC Name 1-pyridin-3-ylethanone
SMILES CC(=O)C1=CN=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A011225G
View Documents
TCI America
A011225G
25 g
Each for $90.86
 
A0112250G
View Documents
TCI America
A0112250G
250 g
Each for $475.97
 
Description

Description

Specifications

Chemical Identifiers

350-03-8
121.139
WEGYGNROSJDEIW-UHFFFAOYSA-N
9589
CC(=O)C1=CN=CC=C1
C7H7NO
MFCD00006396
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
1-pyridin-3-ylethanone

Specifications

350-03-8
220°C
MFCD00006396
2810
WEGYGNROSJDEIW-UHFFFAOYSA-N
1-pyridin-3-ylethanone
9589
≥98.0% (GC)
3-Acetylpyridine
Yellow
C7H7NO
25 g
3-acetylpyridine, 1-pyridin-3-yl ethanone, 3-acetopyridine, 1-3-pyridinyl ethanone, methyl 3-pyridyl ketone, ethanone, 1-3-pyridinyl, ketone, methyl 3-pyridyl, 3-acetyl pyridine, 3-pyridyl methyl ketone, 1-pyridin-3-yl ethan-1-one
CC(=O)C1=CN=CC=C1
121.139
121.14
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3706

RTECSNumber : OB5425000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'