missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-2,2-dimethyl-1-propanol, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 26734-09-8 |
---|---|
Molecular Formula | C5H13NO |
Molecular Weight (g/mol) | 103.17 |
InChI Key | FNVOFDGAASRDQY-UHFFFAOYSA-N |
Synonym | 3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d |
PubChem CID | 117326 |
IUPAC Name | 3-amino-2,2-dimethylpropan-1-ol |
SMILES | CC(C)(CN)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC457430050
|
Thermo Scientific Chemicals
457430050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC457430250
|
Thermo Scientific Chemicals
457430250 |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
26734-09-8 | |
103.17 | |
3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d | |
3-amino-2,2-dimethylpropan-1-ol |
C5H13NO | |
FNVOFDGAASRDQY-UHFFFAOYSA-N | |
117326 | |
CC(C)(CN)CO |
Specifications
26734-09-8 | |
96% min. (GC) | |
5g | |
FNVOFDGAASRDQY-UHFFFAOYSA-N | |
3-amino-2,2-dimethylpropan-1-ol | |
117326 | |
97% |
Authentic | |
C5H13NO | |
3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d | |
CC(C)(CN)CO | |
103.17 | |
103.17 | |
3-Amino-2,2-dimethyl-1-propanol |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 247-948-3
RUO â Research Use Only