missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2217614

 View more versions of this product

Catalog No. AAB2217614


Explore available promotions

Description

Description

3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
Specifications

Specifications

3-Amino-3-(2-nitrophenyl)propionic acid
C9H10N2O4
3-amino-3-2-nitrophenyl propanoic acid, 3-amino-3-2-nitrophenyl propionic acid, 3-amino-3-2-nitro-phenyl-propionic acid, 3-2-nitrophenyl-dl-beta-alanine, 3-2-nitrophenyl-beta-alanine, rs-3-amino-3-2-nitrophenyl-propionic acid, maybridge1_004875, dl-beta-2-nitrophenyl alanine, beta-amino-o-nitrohydrocinnamic acid, hydrocinnamic acid, beta-amino-o-nitro
XXBOYULKNZTOMN-UHFFFAOYSA-N
3-amino-3-(2-nitrophenyl)propanoic acid
210.189
210.19
5678-48-8
MFCD00090356
Slightly soluble in water.
C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-]
25 g
2779217
98%

Chemical Identifiers

5678-48-8
210.189
XXBOYULKNZTOMN-UHFFFAOYSA-N
2779217
C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-]
C9H10N2O4
MFCD00090356
3-amino-3-2-nitrophenyl propanoic acid, 3-amino-3-2-nitrophenyl propionic acid, 3-amino-3-2-nitro-phenyl-propionic acid, 3-2-nitrophenyl-dl-beta-alanine, 3-2-nitrophenyl-beta-alanine, rs-3-amino-3-2-nitrophenyl-propionic acid, maybridge1_004875, dl-beta-2-nitrophenyl alanine, beta-amino-o-nitrohydrocinnamic acid, hydrocinnamic acid, beta-amino-o-nitro
3-amino-3-(2-nitrophenyl)propanoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only