missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals
$188.53 - $1908.67
Chemical Identifiers
CAS | 5678-48-8 |
---|---|
Molecular Formula | C9H10N2O4 |
Molecular Weight (g/mol) | 210.189 |
MDL Number | MFCD00090356 |
InChI Key | XXBOYULKNZTOMN-UHFFFAOYSA-N |
Synonym | 3-amino-3-2-nitrophenyl propanoic acid, 3-amino-3-2-nitrophenyl propionic acid, 3-amino-3-2-nitro-phenyl-propionic acid, 3-2-nitrophenyl-dl-beta-alanine, 3-2-nitrophenyl-beta-alanine, rs-3-amino-3-2-nitrophenyl-propionic acid, maybridge1_004875, dl-beta-2-nitrophenyl alanine, beta-amino-o-nitrohydrocinnamic acid, hydrocinnamic acid, beta-amino-o-nitro |
PubChem CID | 2779217 |
IUPAC Name | 3-amino-3-(2-nitrophenyl)propanoic acid |
SMILES | C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2217606
|
Thermo Scientific Chemicals
B2217606 |
5 g |
Each for $188.53
|
|
|||||
AAB2217614
|
Thermo Scientific Chemicals
B2217614 |
25 g |
Each for $632.64
|
|
|||||
AAB2217622
|
Thermo Scientific Chemicals
B2217622 |
100 g |
Each for $1,908.67
|
|
|||||
Description
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.
Solubility
Slightly soluble in water.
Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
Chemical Identifiers
5678-48-8 | |
210.189 | |
XXBOYULKNZTOMN-UHFFFAOYSA-N | |
2779217 | |
C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-] |
C9H10N2O4 | |
MFCD00090356 | |
3-amino-3-2-nitrophenyl propanoic acid, 3-amino-3-2-nitrophenyl propionic acid, 3-amino-3-2-nitro-phenyl-propionic acid, 3-2-nitrophenyl-dl-beta-alanine, 3-2-nitrophenyl-beta-alanine, rs-3-amino-3-2-nitrophenyl-propionic acid, maybridge1_004875, dl-beta-2-nitrophenyl alanine, beta-amino-o-nitrohydrocinnamic acid, hydrocinnamic acid, beta-amino-o-nitro | |
3-amino-3-(2-nitrophenyl)propanoic acid |
Specifications
5678-48-8 | |
MFCD00090356 | |
Slightly soluble in water. | |
C1=CC=C(C(=C1)C(CC(=O)O)N)[N+](=O)[O-] | |
210.189 | |
210.19 | |
5 g |
C9H10N2O4 | |
3-amino-3-2-nitrophenyl propanoic acid, 3-amino-3-2-nitrophenyl propionic acid, 3-amino-3-2-nitro-phenyl-propionic acid, 3-2-nitrophenyl-dl-beta-alanine, 3-2-nitrophenyl-beta-alanine, rs-3-amino-3-2-nitrophenyl-propionic acid, maybridge1_004875, dl-beta-2-nitrophenyl alanine, beta-amino-o-nitrohydrocinnamic acid, hydrocinnamic acid, beta-amino-o-nitro | |
XXBOYULKNZTOMN-UHFFFAOYSA-N | |
3-amino-3-(2-nitrophenyl)propanoic acid | |
2779217 | |
98% | |
3-Amino-3-(2-nitrophenyl)propionic acid |
Safety and Handling
TSCA : No
RUO – Research Use Only