Learn More
3-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 219301000
Specifications
3-Amino-4-chlorobenzoic acid | |
2840-28-0 | |
100.0 | |
Beige | |
97.5% min. (HPLC) | |
C7H6ClNO2 | |
MFCD00007671 | |
14, 412 | |
DMGFVJVLVZOSOE-UHFFFAOYSA-N | |
3-amino-4-chlorobenzoic acid | |
76092 | |
98% |
98% | |
97.5 | |
214.0°C to 218.0°C | |
Authentic | |
Plastic bottle | |
H2NC6H3(Cl)CO2H | |
100 g | |
benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213 | |
C1=CC(=C(C=C1C(=O)O)N)Cl | |
171.58 | |
171.58 | |
Powder |
Chemical Identifiers
2840-28-0 | |
171.58 | |
DMGFVJVLVZOSOE-UHFFFAOYSA-N | |
76092 | |
C1=CC(=C(C=C1C(=O)O)N)Cl |
C7H6ClNO2 | |
MFCD00007671 | |
benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213 | |
3-amino-4-chlorobenzoic acid |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 220-635-9
TSCA : TSCA
RUO – Research Use Only