Learn More
3-Amino-4-chlorobenzoic acid, 98%, Thermo Scientific™
$147.80 - $147.80
Chemical Identifiers
CAS | 2840-28-0 |
---|---|
Molecular Formula | C7H6ClNO2 |
Molecular Weight (g/mol) | 171.58 |
MDL Number | MFCD00007671 |
InChI Key | DMGFVJVLVZOSOE-UHFFFAOYSA-N |
Synonym | benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213 |
PubChem CID | 76092 |
IUPAC Name | 3-amino-4-chlorobenzoic acid |
SMILES | C1=CC(=C(C=C1C(=O)O)N)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC219301000
|
Thermo Scientific Chemicals
219301000 |
100 g | Plastic bottle |
Each for $147.80
|
|
||||
Chemical Identifiers
2840-28-0 | |
171.58 | |
DMGFVJVLVZOSOE-UHFFFAOYSA-N | |
76092 | |
C1=CC(=C(C=C1C(=O)O)N)Cl |
C7H6ClNO2 | |
MFCD00007671 | |
benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213 | |
3-amino-4-chlorobenzoic acid |
Specifications
2840-28-0 | |
100.0 | |
Beige | |
97.5% min. (HPLC) | |
C7H6ClNO2 | |
MFCD00007671 | |
14, 412 | |
DMGFVJVLVZOSOE-UHFFFAOYSA-N | |
3-amino-4-chlorobenzoic acid | |
76092 | |
98% | |
3-Amino-4-chlorobenzoic acid, 98% |
97.5 | |
214.0°C to 218.0°C | |
Authentic | |
Plastic bottle | |
H2NC6H3(Cl)CO2H | |
100 g | |
benzoic acid, 3-amino-4-chloro, unii-3q8r4430cf, 3-amino-4-chloro benzoic acid, 3-amino-4-chloro-benzoic acid, 4-chloro-3-amino benzoic acid, pubchem8988, acmc-1cmgg, 5-carboxy-2-chloroaniline, dsstox_cid_24414, dsstox_rid_80213 | |
C1=CC(=C(C=C1C(=O)O)N)Cl | |
171.58 | |
171.58 | |
Powder |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 220-635-9
TSCA : TSCA
RUO – Research Use Only