Learn More
3-Amino-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 271490050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Amino-4-hydroxybenzoic acid | |
1571-72-8 | |
100.0 | |
Beige-Gray to Brown | |
97.5% min. (HPLC) | |
C7H7NO3 | |
MFCD00007697 | |
benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504 | |
C1=CC(=C(C=C1C(=O)O)N)O | |
153.14 | |
CHEBI:29476 | |
98% |
98% | |
97.5 | |
198.0°C to 205.0°C | |
Authentic | |
Glass bottle | |
H2NC6H3(OH)COOH | |
5 g | |
MRBKRZAPGUCWOS-UHFFFAOYSA-N | |
3-amino-4-hydroxybenzoic acid | |
65083 | |
153.14 | |
Crystalline Powder |
Chemical Identifiers
1571-72-8 | |
153.14 | |
MRBKRZAPGUCWOS-UHFFFAOYSA-N | |
65083 | |
3-amino-4-hydroxybenzoic acid |
C7H7NO3 | |
MFCD00007697 | |
benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504 | |
CHEBI:29476 | |
C1=CC(=C(C=C1C(=O)O)N)O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Warning
EINECSNumber : 216-390-2
TSCA : TSCA
RUO – Research Use Only