missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals

$72.79 - $267.62

Chemical Identifiers

CAS 1571-72-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007697
InChI Key MRBKRZAPGUCWOS-UHFFFAOYSA-N
Synonym benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
PubChem CID 65083
ChEBI CHEBI:29476
IUPAC Name 3-amino-4-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC271490050
SDS View Documents
Thermo Scientific Chemicals
271490050
5 g
Each for $72.79
 
AC271490250
SDS View Documents
Thermo Scientific Chemicals
271490250
25 g
Each for $267.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1571-72-8
153.14
MRBKRZAPGUCWOS-UHFFFAOYSA-N
65083
3-amino-4-hydroxybenzoic acid
C7H7NO3
MFCD00007697
benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
CHEBI:29476
C1=CC(=C(C=C1C(=O)O)N)O

Specifications

1571-72-8
100.0
Beige-Gray to Brown
97.5% min. (HPLC)
C7H7NO3
MFCD00007697
benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
C1=CC(=C(C=C1C(=O)O)N)O
153.14
CHEBI:29476
98%
3-Amino-4-hydroxybenzoic acid, 98%
97.5
198.0°C to 205.0°C
Authentic
Glass bottle
H2NC6H3(OH)COOH
5 g
MRBKRZAPGUCWOS-UHFFFAOYSA-N
3-amino-4-hydroxybenzoic acid
65083
153.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

GHS Signal Word: Warning

EINECSNumber : 216-390-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only