missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-4-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 271490250

 View more versions of this product

Catalog No. AC271490250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Amino-4-hydroxybenzoic acid
1571-72-8
100.0
Beige-Gray to Brown
97.5% min. (HPLC)
C7H7NO3
MFCD00007697
benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
C1=CC(=C(C=C1C(=O)O)N)O
153.14
CHEBI:29476
98%
98%
97.5
198.0°C to 205.0°C
Authentic
Glass bottle
H2NC6H3(OH)COOH
25 g
MRBKRZAPGUCWOS-UHFFFAOYSA-N
3-amino-4-hydroxybenzoic acid
65083
153.14
Crystalline Powder

Chemical Identifiers

1571-72-8
153.14
MRBKRZAPGUCWOS-UHFFFAOYSA-N
65083
3-amino-4-hydroxybenzoic acid
C7H7NO3
MFCD00007697
benzoic acid, 3-amino-4-hydroxy, cheetan, 3,4-ahba, 3-amino-4-hydroxy-benzoic acid, 5-amino saliciylic acid, 3-amino-4-hydroxy benzoic acid, 3-amino-4-hydroxybenzoic acid hydrochloride, 3-amino-4-hydroxybenzoicacid, 5-amino saliciylic acid 5-asa, pubchem20504
CHEBI:29476
C1=CC(=C(C=C1C(=O)O)N)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

GHS Signal Word: Warning

EINECSNumber : 216-390-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only